Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
MaxiSorp
™
384 well plates
Products
Product Category
plates (3)
Color
Manufacturer
Binding Type
Length (mm)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
P6366
Nunc
®
MaxiSorp
™
384 well plates,
384 well plate, clear, polystyrene, 120ul/well, pinchbar design, 30/cs
Expand
View Pricing
P6491
Nunc
®
MaxiSorp
™
384 well plates,
384 well plate, black, polystyrene, 120ul/well, pinchbar design, 30/cs
Expand
View Pricing
P6116
Nunc
®
MaxiSorp
™
384 well plates,
384 well plate, white, polystyrene, 120ul/well, pinchbar design, 30/cs, size 384 wells, flat bottom white polystyrene wells, maximum volume 120 μL
Expand
View Pricing