Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
Edge 2.0 96-well Plate
Products
Product Category
plates (10)
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
TMO167574
Nunc
®
Edge 2.0 96-well Plate,
Nunclon
®
Delta surface, polystyrene, sterile, without lid, clear, case of 50 ea
Expand
View Pricing
TMO267427
Nunc
®
Edge 2.0 96-well Plate,
untreated, polystyrene, sterile, with lid, clear, case of 50 ea
Expand
View Pricing
TMO267544
Nunc
®
Edge 2.0 96-well Plate,
untreated, polystyrene, sterile, with lid, clear, case of 160 ea
Expand
View Pricing
TMO167425
Nunc
®
Edge 2.0 96-well Plate,
Nunclon
®
Delta surface, polystyrene, sterile, with lid, clear, case of 50 ea
Expand
View Pricing
TMO167542
Nunc
®
Edge 2.0 96-well Plate,
Nunclon
®
Delta surface, polystyrene, sterile, with lid, clear, case of 160 ea
Expand
View Pricing
TMO267566
Nunc
®
Edge 2.0 96-well Plate,
untreated, polystyrene, non-sterile, without lid, clear, case of 50 ea
Expand
View Pricing
TMO267576
Nunc
®
Edge 2.0 96-well Plate,
untreated, polystyrene, sterile, without lid, clear, case of 50 ea
Expand
View Pricing
TMO167554
Nunc
®
Edge 2.0 96-well Plate,
Nunclon
®
Delta surface, polystyrene, sterile, without lid, clear, case of 160 ea
Expand
View Pricing
TMO267556
Nunc
®
Edge 2.0 96-well Plate,
untreated, polystyrene, sterile, without lid, clear, case of 160 ea
Expand
View Pricing
TMO267578
Nunc
®
Edge 2.0 96-well Plate,
untreated, polystyrene, non-sterile, with lid, clear, case of 160 ea
Expand
View Pricing