
N-Sulfocarbamoylsaxitoxin solution

Synonym(s):
GTX5 solution, Gonyautoxin solution, Toxin B1 solution
Empirical Formula (Hill Notation):
C10H17N7O7S
CAS Number:
Molecular Weight:
379.35
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing