
Neochlorogenic acid

Synonym(s):
5-O-(trans-3,4-Dihydroxycinnamoyl)-D-quinic acid, trans-5-O-Caffeoylquinic acid
Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing