
Nefazodone hydrochloride

Synonym(s):
2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl- 2,4-dihydro-4-(2-phenoxyethyl)-3H-1,2,4-triazol-3-one hydrochloride, BMY-13754, MJ-13754-1, Serzone
Empirical Formula (Hill Notation):
C25H32ClN5O2 · HCl
CAS Number:
Molecular Weight:
506.47
Products
Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2