
Naphthol Green B

Synonym(s):
Acid Green 1
Empirical Formula (Hill Notation):
C30H15FeN3Na3O15S3
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing