Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
NanoFabTx
™
device accessory
Products
Product Category
tubing (2)
filters (1)
Brand
Sigma-Aldrich (6)
Quality Segment
Application
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
916609
NanoFabTx
™
device accessory,
end fittings for 1.6 mm tubings
Expand
View Pricing
917230
NanoFabTx
™
device accessory,
interface H
Expand
View Pricing
917877
NanoFabTx
™
device accessory,
in line filter
Expand
View Pricing
917494
NanoFabTx
™
device accessory,
tubing, 250 μm FEP, 10 m, with ferrules
Expand
View Pricing
916862
NanoFabTx
™
device accessory,
tubing, 100 μm FEP, 10 m, with ferrules
Expand
View Pricing
916986
NanoFabTx
™
device accessory,
top interface
Expand
View Pricing