
Nabumetone Related Compound A

Synonym(s):
(E)-4-(6-Methoxynaphthalen-2-yl)but-3-en-2-one
Empirical Formula (Hill Notation):
C15H14O2
CAS Number:
Molecular Weight:
226.27
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing