
MPTP Hydrochloride

Synonym(s):
MPTP hydrochloride
Empirical Formula (Hill Notation):
C12H15N · HCl
CAS Number:
Molecular Weight:
209.72
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing