Skip to Content
Merck
Monoclonal Anti-TUBB3 antibody produced in mouse clone TU-20, purified immunoglobulin, buffered aqueous solution

Monoclonal Anti-TUBB3 antibody produced in mouse

Monoclonal Anti-TUBB3 antibody produced in mouse clone TU-20, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-βIII-tubulin

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique