Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molybdenum-rhenium alloy, Mo52.5Re47.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GF82109482
Molybdenum-rhenium alloy, Mo52.5Re47.5,
1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF14023156
Molybdenum-rhenium alloy, Mo52.5Re47.5,
foil, 50 x 50mm, 0.050mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF60768580
Molybdenum-rhenium alloy, Mo52.5Re47.5,
5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF67856877
Molybdenum-rhenium alloy, Mo52.5Re47.5,
0.5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF11863481
Molybdenum-rhenium alloy, Mo52.5Re47.5,
foil, 50 x 50mm, 0.125mm thickness, annealed
Expand
View Pricing
GF52894907
Molybdenum-rhenium alloy, Mo52.5Re47.5,
foil, 25 x 25mm, 0.125mm thickness, annealed
Expand
View Pricing
GF09142398
Molybdenum-rhenium alloy, Mo52.5Re47.5,
0.5m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF82935074
Molybdenum-rhenium alloy, Mo52.5Re47.5,
foil, 100 x 100mm, 0.125mm thickness, annealed
Expand
View Pricing
GF49964754
Molybdenum-rhenium alloy, Mo52.5Re47.5,
2m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF62942856
Molybdenum-rhenium alloy, Mo52.5Re47.5,
10m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF66884125
Molybdenum-rhenium alloy, Mo52.5Re47.5,
1m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF42799719
Molybdenum-rhenium alloy, Mo52.5Re47.5,
1m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF96073139
Molybdenum-rhenium alloy, Mo52.5Re47.5,
0.2m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF60516020
Molybdenum-rhenium alloy, Mo52.5Re47.5,
2m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF63686144
Molybdenum-rhenium alloy, Mo52.5Re47.5,
2m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF42512183
Molybdenum-rhenium alloy, Mo52.5Re47.5,
foil, 25 x 25mm, 0.050mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF64503547
Molybdenum-rhenium alloy, Mo52.5Re47.5,
5m wire, 0.125mm diameter, as drawn
Expand
View Pricing