
Misoprostol for system suitability

Synonym(s):
Misoprostol, (±)-15-Deoxy-(16RS)-16-hydroxy-16-methylprostaglandin E1 methyl ester
Empirical Formula (Hill Notation):
C22H38O5
CAS Number:
Molecular Weight:
382.53
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing