
Midodrine Related Compound A

Synonym(s):
2-Amino-1-(2,5-dimethoxyphenyl)ethanol, Desglymidodrine
Empirical Formula (Hill Notation):
C10H15NO3
CAS Number:
Molecular Weight:
197.23
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2