Skip to Content
Merck
Methyl trimethylsilyl dimethylketene acetal 95%

Methyl trimethylsilyl dimethylketene acetal

Methyl trimethylsilyl dimethylketene acetal 95%
Synonym(s):
1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, (1-Methoxy-2-methyl-1-propenyloxy)trimethylsilane, MTDA
Linear Formula:
(CH3)2C=C(OCH3)OSi(CH3)3
CAS Number:
Molecular Weight:
174.31

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing