Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyl Red sodium salt
Synonym(s):
4-Dimethylaminoazobenzene-2′-carboxylic acid sodium salt, Acid Red 2
Empirical Formula (Hill Notation):
C
15
H
14
N
3
NaO
2
CAS Number:
845-10-3
Molecular Weight:
291.28
EC Number:
212-682-9
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (2)
Formula Weight
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
114502
Methyl Red sodium salt,
ACS reagent, Dye content 95 %
Expand
View Pricing
M8256
Methyl Red sodium salt,
Powder
Expand
View Pricing