Skip to Content
Products
Cart
0
AN
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Methyl
cis
-11-eicosenoate
Synonym(s):
cis
-11-Eicosenoic acid methyl ester
Linear Formula:
CH
3
(CH
2
)
7
HC=CH(CH
2
)
9
CO
2
CH
3
CAS Number:
2390-09-2
Molecular Weight:
324.54
EC Number:
219-226-8
Products
Product Category
lipids (2)
fatty acids (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
E6885
Methyl
cis
-11-eicosenoate,
≥98%
Expand
View Pricing
17263
Methyl
cis
-11-eicosenoate,
analytical standard
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More