Methyl 5-methoxy-3-oxovalerate
Synonym(s):
Methyl 5-methoxy-3-oxopentanoate, Nazarov reagent
Linear Formula:
CH3O(CH2)2COCH2COOCH3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing