Skip to Content
Merck
Mercury(II) acetate puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)

Mercury(II) acetate

Mercury(II) acetate puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)
Synonym(s):
Mercuric acetate, Mercury(II) acetate
Linear Formula:
Hg(CH3COO)2
CAS Number:
Molecular Weight:
318.68

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade