Skip to Content
Merck
Menaquinone (K2) analytical standard

Menaquinone (K2)

Menaquinone (K2) analytical standard
Synonym(s):
Vitamin K2, Menaquinone 4, Menatetrenone
Empirical Formula (Hill Notation):
C31H40O2
CAS Number:
Molecular Weight:
444.65

Products

Product Category
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing