Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lead(II) citrate tribasic trihydrate
Synonym(s):
Citric acid lead salt
Linear Formula:
(C
6
H
5
O
7
)
2
Pb
3
· 3H
2
O
CAS Number:
6107-83-1
Molecular Weight:
1053.85
EC Number:
208-141-1
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
15326
Lead(II) citrate tribasic trihydrate,
purum, suitable for electron microscopy
Expand
View Pricing