Skip to Content
Merck
L-Dihydroorotic acid ≥99%

L-Dihydroorotic acid

L-Dihydroorotic acid ≥99%
Synonym(s):
2,6-Dioxohexahydro-4-pyrimidinecarboxylic acid, L-Hydroorotic acid, Dihydro-L-orotic acid
Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing