Skip to Content
Merck
L-Cystine dihydrochloride EMPROVE® EVOLVE

L-Cystine dihydrochloride

L-Cystine dihydrochloride EMPROVE® EVOLVE
Empirical Formula (Hill Notation):
C6H12N2O4S2 · 2HCl
CAS Number:
Molecular Weight:
313.22

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Optical Isomer
Amino Acid
Technique
Showing 1-3 of 3