Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Julabo
®
cryo-compact circulator
Products
Product Category
baths (4)
Manufacturer
Capacity (L)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z743138
Julabo
®
cryo-compact circulator,
CF31, capacity 3.5 L, temperature -30-200 °C, precision: ±0.02, AC/DC input 115 V AC, US 3-pin plug (6-20p)
Expand
View Pricing
Z743141
Julabo
®
cryo-compact circulator,
CF41, capacity 5.5 L, temperature -40-200 °C, precision: ±0.02, AC/DC input 115 V AC, US 3-pin plug (6-20p)
Expand
View Pricing
Z743131
Julabo
®
cryo-compact circulator,
CF30, capacity 3.5 L, temperature -30-150 °C, precision: ±0.03, AC/DC input 115 V AC, US 3-pin plug (5-15p)
Expand
View Pricing
Z743134
Julabo
®
cryo-compact circulator,
CF40, capacity 5.5 L, temperature -40-150 °C, precision: ±0.03, AC/DC input 115 V AC, US 3-pin plug (5-15p)
Expand
View Pricing