
Isoeugenyl acetate

Synonym(s):
NSC 46121
Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3