Skip to Content
Merck
Insulin chain A oxidized ammonium salt from bovine pancreas ≥80% (HPLC), powder

Insulin chain A oxidized ammonium salt from bovine pancreas

Insulin chain A oxidized ammonium salt from bovine pancreas ≥80% (HPLC), powder
Linear Formula:
C97H151N25O46S4 · (NH3)x
CAS Number:
Molecular Weight:
2531.64 (free acid basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Sterility
Showing 1-1 of 1