
Guaifenesin Impurity B

Synonym(s):
2-(2-Methoxyphenoxy)-1,3-propanediol, 2-(2-Methoxyphenoxy)propane-1,3-diol
Empirical Formula (Hill Notation):
C10H14O4
CAS Number:
Molecular Weight:
198.22
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing