Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glass Sealing Alloy, Fe54Ni29Co17
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF40100050
Glass Sealing Alloy, Fe54Ni29Co17,
100mm rod, 2.0mm diameter
Expand
View Pricing
GF94404371
Glass Sealing Alloy, Fe54Ni29Co17,
500mm rod, 3.2mm diameter
Expand
View Pricing
GF64040239
Glass Sealing Alloy, Fe54Ni29Co17,
100mm tube, 6.35mm outside diameter, 0.25mm wall thickness, 5.85mm inside diameter, as drawn
Expand
View Pricing
GF73818850
Glass Sealing Alloy, Fe54Ni29Co17,
100mm tube, 6.0mm outside diameter, 0.5mm wall thickness, 5mm inside diameter, as drawn
Expand
View Pricing
GF71024420
Glass Sealing Alloy, Fe54Ni29Co17,
foil, 15mm disks, 0.20mm thickness, annealed
Expand
View Pricing
GF12842737
Glass Sealing Alloy, Fe54Ni29Co17,
foil, 50 x 50mm, 0.20mm thickness, annealed
Expand
View Pricing
GF56271078
Glass Sealing Alloy, Fe54Ni29Co17,
100mm rod, 3.2mm diameter
Expand
View Pricing
GF07753091
Glass Sealing Alloy, Fe54Ni29Co17,
200mm rod, 1.6mm diameter
Expand
View Pricing
GF89993952
Glass Sealing Alloy, Fe54Ni29Co17,
100mm rod, 1.6mm diameter
Expand
View Pricing
GF59889043
Glass Sealing Alloy, Fe54Ni29Co17,
100mm rod, 1.25mm diameter
Expand
View Pricing
GF07433793
Glass Sealing Alloy, Fe54Ni29Co17,
200mm tube, 4.24mm outside diameter, 0.38mm wall thickness, 3.48mm inside diameter, as drawn
Expand
View Pricing
GF29659472
Glass Sealing Alloy, Fe54Ni29Co17,
100mm tube, 4.24mm outside diameter, 0.38mm wall thickness, 3.48mm inside diameter, as drawn
Expand
View Pricing
GF08037592
Glass Sealing Alloy, Fe54Ni29Co17,
100mm tube, 2.0mm outside diameter, 0.5mm wall thickness, 1.0mm inside diameter, as drawn
Expand
View Pricing
GF09484645
Glass Sealing Alloy, Fe54Ni29Co17,
200mm tube, 0.6mm outside diameter, 0.1mm wall thickness, 0.4mm inside diameter, as drawn
Expand
View Pricing
GF35542807
Glass Sealing Alloy, Fe54Ni29Co17,
foil, 150 x 150mm, 0.25mm thickness, annealed
Expand
View Pricing
GF40271071
Glass Sealing Alloy, Fe54Ni29Co17,
foil, 50 x 50mm, 0.25mm thickness, annealed
Expand
View Pricing
GF99782993
Glass Sealing Alloy, Fe54Ni29Co17,
foil, 50 x 50mm, 0.025mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF64798262
Glass Sealing Alloy, Fe54Ni29Co17,
foil, 25 x 25mm, 0.025mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF87308501
Glass Sealing Alloy, Fe54Ni29Co17,
foil, 50 x 50mm, 0.125mm thickness, annealed
Expand
View Pricing
GF09308875
Glass Sealing Alloy, Fe54Ni29Co17,
50mm rod, 19mm diameter
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
Page 1 of 11
Page 1 of 11
English - EN
Learn More