
Gatifloxacin sesquihydrate

Synonym(s):
Gatifloxacin sesquihydrate, (±)-1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid sesquihydrate
Empirical Formula (Hill Notation):
C19H22FN3O4 · 1.5H2O
CAS Number:
Molecular Weight:
402.42
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing