Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fusaric acid
Synonym(s):
5-Butylpicolinic acid, 5-Butylpyridine-2-carboxylic acid
Empirical Formula (Hill Notation):
C
10
H
13
NO
2
CAS Number:
536-69-6
Molecular Weight:
179.22
EC Number:
208-643-0
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
F6513
Fusaric acid,
from
Gibberella fujikuroi
Expand
View Pricing
55952
Fusaric acid,
for HPLC derivatization, ≥99.0% (HPLC)
Expand
View Pricing