Skip to Content
Merck
Formamidinium bromide ≥98%

Formamidinium bromide

Formamidinium bromide ≥98%
Empirical Formula (Hill Notation):
CH5BrN2
CAS Number:
Molecular Weight:
124.97

Products

Product Category
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing