
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate

Synonym(s):
TFFH
Linear Formula:
[FC[=N(CH3)2]N(CH3)2]PF6
CAS Number:
Molecular Weight:
264.12
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Application
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing