
Fludrocortisone acetate

Synonym(s):
9α-Fluoro-11β,17α,21-trihydroxy-4-pregnene-3,20-dione acetate, 9-Fluorocortisol acetate, 9-Fluorohydrocortisone acetate
Empirical Formula (Hill Notation):
C23H31FO6
CAS Number:
Molecular Weight:
422.49
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Sterility
Showing 1-4 of 4
Compare
Product Number
Description
Pricing