Skip to Content
Merck
Fast Blue B Salt Dye content ~95 %

Fast Blue B Salt

Fast Blue B Salt Dye content ~95 %
Synonym(s):
o-Dianisidine bis(diazotized) zinc double salt, Azoic Diazo No. 48, DBB, Diazo Blue B, Naphthanil Diazo Blue B
Linear Formula:
C14H12N4O2Cl2 · ZnCl2
CAS Number:
Molecular Weight:
475.47
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing