Skip to Content
Merck
Ethylenediaminetetraacetic acid disodium salt solution

Ethylenediaminetetraacetic acid disodium salt solution

Ethylenediaminetetraacetic acid disodium salt solution
Synonym(s):
(Ethylenedinitrilo)tetraacetic acid disodium salt, EDTA disodium salt, EDTA-Na2
Linear Formula:
[-CH2N(CH2CO2Na)CH2CO2H]2
CAS Number:
Molecular Weight:
336.21