
Ethosuximide Related Compound A

Synonym(s):
2-Ethyl-2-methylsuccinic acid
Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing