Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers
Products
Product Category
thermocyclers (9)
Manufacturer
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
EP6331000025
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
Gradient, complete unit, aluminum block, AC/DC input 115 V, 50/60 Hz (US)
Expand
View Pricing
EP6336000023
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
GX2, AC/DC input 110 V, 50 - 60 Hz
Expand
View Pricing
EP6333000022
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
Nexus, complete unit, aluminum block, AC/DC input 115 V, 50/60 Hz (US)
Expand
View Pricing
EP6332000029
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
Eco, satellite unit, aluminum block, AC/DC input 115 V, 50/60 Hz (US)
Expand
View Pricing
EP6334000026
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
Gradient Eco, Eco units without control panels, AC/DC input 110 V, 50 - 60 Hz
Expand
View Pricing
EP6346000021
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
SX1, AC/DC input 110 V, 50 - 60 Hz
Expand
View Pricing
EP6339000024
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
X2e, Eco units without control panels, AC/DC input 110 V, 50 - 60 Hz
Expand
View Pricing
EP6348000029
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
SX1e, Eco units without control panels, AC/DC input 110 V, 50 - 60 Hz
Expand
View Pricing
EP6338000020
Eppendorf
®
Mastercycler
®
Nexus Thermal Cyclers,
GX2e, Eco units without control panels, AC/DC input 110 V, 50 - 60 Hz
Expand
View Pricing