Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
Sterile, Racks
Products
Product Category
pipette tips (6)
Manufacturer
Volume (ml)
Volume (µL)
Color
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP0030071557
Eppendorf
®
epT.I.P.S.
®
Sterile, Racks,
volume 0.1-20 μL, sterile, pkg of 960 ea (10 x racks of 96ea)
Expand
View Pricing
EP0030071581
Eppendorf
®
epT.I.P.S.
®
Sterile, Racks,
volume 50-1,000 μL, sterile, pkg of 960 ea (10 x racks of 96ea)
Expand
View Pricing
EP0030071565
Eppendorf
®
epT.I.P.S.
®
Sterile, Racks,
volume 2-200 μL, sterile, pkg of 960 ea (10 x racks of 96ea)
Expand
View Pricing
EP0030071573
Eppendorf
®
epT.I.P.S.
®
Sterile, Racks,
volume 20-300 μL, sterile, pkg of 960 ea (10 x racks of 96ea)
Expand
View Pricing
EP0030071620
Eppendorf
®
epT.I.P.S.
®
Sterile, Racks,
volume 0.1-5 mL, sterile, pkg of 960 ea (10 x racks of 96ea)
Expand
View Pricing
EP0030071590
Eppendorf
®
epT.I.P.S.
®
Sterile, Racks,
volume 50-1,250 μL, sterile, pkg of 960 ea (10 x racks of 96ea)
Expand
View Pricing