Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
ep Dualfilter T.I.P.S.
®
Forensic DNA Grade
Products
Product Category
pipette tips (4)
Manufacturer
Volume (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP0030078810
Eppendorf
®
ep Dualfilter T.I.P.S.
®
Forensic DNA Grade,
tip volume × L 0.1-10 μL × 40 mm, gray tip, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078837
Eppendorf
®
ep Dualfilter T.I.P.S.
®
Forensic DNA Grade,
tip volume × L 2-200 μL × 55 mm, yellow tip, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078845
Eppendorf
®
ep Dualfilter T.I.P.S.
®
Forensic DNA Grade,
tip volume × L 50-1000 μL × 76 mm, blue tip, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078829
Eppendorf
®
ep Dualfilter T.I.P.S.
®
Forensic DNA Grade,
tip volume × L 2-20 μL × 53 mm, yellow tip, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing