
Emamectin Benzoate B1a

Empirical Formula (Hill Notation):
C56H81NO15
CAS Number:
Molecular Weight:
1008.24
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing