Skip to Content
Merck
ECO TWEEN® 20 viscous liquid

ECO TWEEN® 20

ECO TWEEN® 20 viscous liquid
Synonym(s):
TWEEN® 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate
CAS Number:

Products

Product Category
Sustainability
Feature
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing