
Dutasteride

Synonym(s):
(5α,17β)-N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-azaandrost-1-ene-17-carboxamide
Empirical Formula (Hill Notation):
C27H30F6N2O2
CAS Number:
Molecular Weight:
528.53
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3