Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
laboratory bottles, with caps
Products
Product Category
bottles (19)
Feature
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Package Size
Sterility
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
Z305170
Duran
®
laboratory bottles, with caps,
capacity 100 mL, blue PP screw cap and pouring ring
Expand
View Pricing
Z305189
Duran
®
laboratory bottles, with caps,
capacity 250 mL, blue PP screw cap and pouring ring
Expand
View Pricing
Z305200
Duran
®
laboratory bottles, with caps,
capacity 1,000 mL, blue PP screw cap and pouring ring
Expand
View Pricing
Z305197
Duran
®
laboratory bottles, with caps,
capacity 500 mL, blue PP screw cap and pouring ring
Expand
View Pricing
DWK218011453
Duran
®
laboratory bottles, with caps,
bottle capacity 25mL, clear, graduated, GL 25 thread, pouring ring, borosilicate glass 3.3 bottle
Expand
View Pricing
DWK218012955
Duran
®
laboratory bottles, with caps,
bottle capacity 150mL, clear, graduated, GL 45 thread, pouring ring, borosilicate glass 3.3 bottle
Expand
View Pricing
DWK218015155
Duran
®
laboratory bottles, with caps,
bottle capacity 750mL, clear, graduated, GL 45 thread, pouring ring, borosilicate glass 3.3 bottle
Expand
View Pricing
DWK218017353
Duran
®
laboratory bottles, with caps,
capacity 5000 mL, clear, graduated, GL 45 thread, pouring ring, borosilicate glass 3.3 bottle
Expand
View Pricing
DWK218018658
Duran
®
laboratory bottles, with caps,
bottle capacity 10L, clear, graduated, GL 45 thread, pouring ring, borosilicate glass 3.3 bottle
Expand
View Pricing
DWK218016957
Duran
®
laboratory bottles, with caps,
bottle capacity 3500mL, clear, graduated, GL 45 thread, pouring ring, borosilicate glass 3.3 bottle
Expand
View Pricing
DWK218019157
Duran
®
laboratory bottles, with caps,
bottle capacity 20L, clear, graduated, GL 45 thread, pouring ring, borosilicate glass 3.3 bottle
Expand
View Pricing
Z305219
Duran
®
laboratory bottles, with caps,
capacity 2,000 mL, blue PP screw cap and pouring ring
Expand
View Pricing
Z305162
Duran
®
laboratory bottles, with caps,
capacity 50 mL, blue PP screw cap and pouring ring
Expand
View Pricing
DWK218011753
Duran
®
laboratory bottles, with caps,
clear, graduated, neck joint: threaded GL32, pouring ring, bottle capacity 50 mL
Expand
View Pricing
DWK218012458
Duran
®
laboratory bottles, with caps,
clear, graduated, neck joint: threaded GL45, pouring ring, bottle capacity 100 mL
Expand
View Pricing
DWK218013651
Duran
®
laboratory bottles, with caps,
clear, graduated, neck joint: threaded GL45, pouring ring, bottle capacity 250 mL
Expand
View Pricing
DWK218014459
Duran
®
laboratory bottles, with caps,
clear, graduated, neck joint: threaded GL45, pouring ring, bottle capacity 500 mL
Expand
View Pricing
DWK218015455
Duran
®
laboratory bottles, with caps,
clear, graduated, neck joint: threaded GL45, pouring ring, bottle capacity 1000 mL
Expand
View Pricing
DWK218016357
Duran
®
laboratory bottles, with caps,
clear, graduated, neck joint: threaded GL45, pouring ring, bottle capacity 2000 mL
Expand
View Pricing