Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DL-Dithiothreitol
Synonym(s):
threo
-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT, DL-Dithiothreitol
Linear Formula:
HSCH
2
CH(OH)CH(OH)CH
2
SH
CAS Number:
3483-12-3
Molecular Weight:
154.25
EC Number:
222-468-7
Products
Brand
SAFC (1)
Sigma-Aldrich (7)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reagent Type
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
D9779
DL-Dithiothreitol,
for molecular biology, ≥98% (HPLC), ≥99% (titration)
Expand
View Pricing
43815
DL-Dithiothreitol,
BioUltra, for molecular biology, ≥99.5% (RT)
Expand
View Pricing
43819
DL-Dithiothreitol,
≥99.0% (RT)
Expand
View Pricing
D9163
DL-Dithiothreitol,
suitable for electrophoresis, ≥99%
Expand
View Pricing
D0632
DL-Dithiothreitol,
≥98% (HPLC), ≥99.0% (titration)
Expand
View Pricing
1.37159
DL-Dithiothreitol,
EMPROVE
®
ESSENTIAL
Expand
View Pricing
D5545
DL-Dithiothreitol,
BioXtra, ≥99.0% (titration)
Expand
View Pricing
07-6130
DL-Dithiothreitol,
≥99.0%
Expand
View Pricing