![Diethyl phthalate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/225/112/b50c3311-73eb-45e3-86be-389921ea8558/640/b50c3311-73eb-45e3-86be-389921ea8558.png)
Diethyl phthalate
![Diethyl phthalate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/225/112/b50c3311-73eb-45e3-86be-389921ea8558/640/b50c3311-73eb-45e3-86be-389921ea8558.png)
Synonym(s):
Diethyl phthalate
Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing