
Cyanocobalamin for system suitability

Synonym(s):
Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin
Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing