Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cryo-Tags
®
labels
Products
Product Category
labels (6)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Length (in)
Color
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z376914
Cryo-Tags
®
labels,
white, W × L 0.75 in. × 1.5 in., pkg of 1000 ea per roll
Expand
View Pricing
Z743350
Cryo-Tags
®
labels,
orange, L × W 1.50 in. × 0.75 in., pkg of 1000 ea per roll
Expand
View Pricing
Z743347
Cryo-Tags
®
labels,
blue, L × W 1.50 in. × 0.75 in., pkg of 1000 ea per roll
Expand
View Pricing
Z743349
Cryo-Tags
®
labels,
green, L × W 1.50 in. × 0.75 in., pkg of 1000 ea per roll
Expand
View Pricing
Z743352
Cryo-Tags
®
labels,
yellow, L × W 1.50 in. × 0.75 in., pkg of 1000 ea per roll
Expand
View Pricing
Z743351
Cryo-Tags
®
labels,
red, L × W 1.50 in. × 0.75 in., pkg of 1000 ea per roll
Expand
View Pricing