Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Snap-Seal plastic sample containers
Products
Manufacturer
Diameter (mm)
Volume (ml)
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS173010
Corning
®
Snap-Seal plastic sample containers,
volume 300 mL, diam. × H 63 mm × 112 mm
Expand
View Pricing
CLS17302C
Corning
®
Snap-Seal plastic sample containers,
volume 45 mL, diam. × H 30 mm × 84 mm
Expand
View Pricing
CLS17304H
Corning
®
Snap-Seal plastic sample containers,
volume 120 mL
Expand
View Pricing
CLS17304L
Corning
®
Snap-Seal plastic sample containers,
volume 120 mL, diam. × H 68 mm × 52 mm
Expand
View Pricing
CLS17308
Corning
®
Snap-Seal plastic sample containers,
volume 240 mL, diam. × H 80 mm × 75 mm
Expand
View Pricing