Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Costar
®
low binding microcentrifuge tubes
Products
Product Category
tubes (2)
Color
Manufacturer
Volume (ml)
Package Size
Special Grade
Natural (1)
Sterility
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
CLS3207
Corning
®
Costar
®
low binding microcentrifuge tubes,
1.7mL low binding snap cap microcentrifuge tube, polypropylene, non-sterile, 250/cs
Expand
View Pricing
CLS3206
Corning
®
Costar
®
low binding microcentrifuge tubes,
0.65mL low binding snap cap microcentrifuge tube, polypropylene, non-sterile, natural, 500/cs
Expand
View Pricing