Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper
Empirical Formula (Hill Notation):
Cu
Molecular Weight:
63.55
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
807958
Copper,
nanowires, diam. × L ~100 nm (±20 nm) × 10~20 μm, ~5 mg/mL in ethanol
Expand
View Pricing
807842
Copper,
nanowires, diam. × L ~100 nm (±20 nm) × 10~20 μm, ~5 mg/mL (in IPA)
Expand
View Pricing
808040
Copper,
nanowires, diam. × L ~100 nm (±20 nm) × 10~20 μm, Wet cake (Wet cake)
Expand
View Pricing