Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
GF14725830
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
100m wire, 0.25mm diameter, hard
Expand
View Pricing
GF46551944
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
100m wire, 0.5mm diameter, hard
Expand
View Pricing
GF04523475
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
10m wire, 1.0mm diameter, hard
Expand
View Pricing
GF02622349
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
20m wire, 0.5mm diameter, hard
Expand
View Pricing
GF24194774
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
100m wire, 1.0mm diameter, hard
Expand
View Pricing
GF06001603
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
5m wire, 1.0mm diameter, hard
Expand
View Pricing
GF20136811
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
200m wire, 0.5mm diameter, hard
Expand
View Pricing
GF74121892
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
50m wire, 0.25mm diameter, hard
Expand
View Pricing
GF20548515
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
200m wire, 0.25mm diameter, hard
Expand
View Pricing
GF81044872
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
20m wire, 0.25mm diameter, hard
Expand
View Pricing
GF94383591
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
50m wire, 1.0mm diameter, hard
Expand
View Pricing
GF07528079
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
50m wire, 0.5mm diameter, hard
Expand
View Pricing
GF28425078
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
10m wire, 0.5mm diameter, hard
Expand
View Pricing
GF43275584
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
500m wire, 0.25mm diameter, hard
Expand
View Pricing
GF51471446
Cobalt-chromium-iron-nickel-molybdenum-manganese alloy, Co40Cr20Fe15Ni15Mo7Mn2CBe,
20m wire, 1.0mm diameter, hard
Expand
View Pricing